siRNA Details
| Gene Symbol: ZDHHC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318015.1 | NP_001304944.1 | 5 | 1130 | ||
| NM_001318016.1 | NP_001304945.1 | 5 | 993 | |||
| NM_001318020.1 | NP_001304949.1 | 5 | 943 | |||
| NM_001318023.1 | NP_001304952.1 | 5 | 962 | |||
| NM_032799.4 | NP_116188.2 | 5 | 965 | |||
| XM_011519116.2 | XP_011517418.1 | 5 | 1117 | |||
| GenBank mRNA | AK027430.1 | - | 6 | 802 | ||
| AK075332.1 | - | 6 | 802 | |||
| AK092843.1 | - | 4 | 2360 | |||
| BC009280.2 | - | 5 | 942 | 4302257 | ||
| BC048251.1 | - | 5 | 1111 | 5418875 | ||

ZDHHC12
Genomic Map