siRNA Details
| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032800.2 | NP_116189.1 | 1 | 143 | ||
| GenBank mRNA | AK027431.1 | - | 1 | 118 | ||
| BC066649.1 | - | 1 | 131 | 6011914 | ||
| DA754292.1 | - | 1 | 143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf198| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032800.2 | NP_116189.1 | 1 | 143 | ||
| GenBank mRNA | AK027431.1 | - | 1 | 118 | ||
| BC066649.1 | - | 1 | 131 | 6011914 | ||
| DA754292.1 | - | 1 | 143 | |||