siRNA Details
| Gene Symbol: C1orf198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136494.1 | NP_001129966.1 | 5, 6 | 1032 | ||
| NM_001136495.1 | NP_001129967.1 | 3, 4 | 756 | |||
| NM_032800.2 | NP_116189.1 | 3, 4 | 974 | |||
| XM_017002599.1 | XP_016858088.1 | 4, 5 | 1239 | |||
| GenBank mRNA | AK027431.1 | - | 3, 4 | 949 | ||
| AK096166.1 | - | 5, 6 | 1032 | |||
| AK131306.1 | - | 3, 4 | 883 | |||
| AK292433.1 | - | 3, 4 | 756 | |||
| AL713769.1 | - | 3, 4 | 723 | |||
| BC066649.1 | - | 3, 4 | 962 | 6011914 | ||

C1orf198
Genomic Map