siRNA Details
| Gene Symbol: MIGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032809.2 | NP_116198.2 | 15 | 1671 | ||
| XM_005252282.1 | XP_005252339.1 | 15 | 1690 | |||
| XM_011519117.1 | XP_011517419.1 | 15 | 1711 | |||
| XM_011519118.1 | XP_011517420.1 | 15 | 1620 | |||
| GenBank mRNA | AK027502.1 | - | 7 | 565 | ||
| AK054974.1 | - | 1 | 617 | |||
| AK074127.1 | - | 16 | 2430 | |||
| AK075421.1 | - | 13 | 1683 | |||
| BC009114.1 | - | 15 | 1671 | 4153683 | ||

MIGA2
Genomic Map