siRNA Details
| Gene Symbol: MIGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032809.2 | NP_116198.2 | 3 | 478 | ||
| XM_005252282.1 | XP_005252339.1 | 3 | 497 | |||
| XM_011519117.1 | XP_011517419.1 | 3 | 518 | |||
| GenBank mRNA | AK074127.1 | - | 3 | 473 | ||
| AK075421.1 | - | 3 | 471 | |||
| AK301241.1 | - | 3 | 467 | |||
| AK303036.1 | - | 3 | 518 | |||
| BC009114.1 | - | 3 | 478 | 4153683 | ||

MIGA2
Genomic Map