siRNA Details
| Gene Symbol: CEP89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032816.4 | NP_116205.3 | 3 | 354 | ||
| XM_005259344.3 | XP_005259401.1 | 3 | 349 | |||
| XM_017027398.1 | XP_016882887.1 | 3 | 349 | |||
| GenBank mRNA | AL832158.1 | - | 3 | 281 | ||
| BC020195.1 | - | 2 | 113 | |||
| BC032307.1 | - | 3 | 288 | |||
| BC136328.1 | - | 3 | 269 | 9020315 | ||
| DA040903.1 | - | 3 | 354 | |||

CEP89
Genomic Map