siRNA Details
| Gene Symbol: CEP89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032816.4 | NP_116205.3 | 14 | 1693 | ||
| XM_005259344.3 | XP_005259401.1 | 14 | 1616 | |||
| XM_011527425.2 | XP_011525727.1 | 11 | 1140 | |||
| XM_017027398.1 | XP_016882887.1 | 14 | 1688 | |||
| GenBank mRNA | AK027546.1 | - | 13 | 1394 | ||
| AL832158.1 | - | 13 | 1516 | |||
| BC136328.1 | - | 14 | 1608 | 9020315 | ||
| BU196203.1 | - | 6 | 670 | |||

CEP89
Genomic Map