siRNA Details
| Gene Symbol: TMEM87B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032824.2 | NP_116213.1 | 3 | 626 | ||
| XM_005263827.2 | XP_005263884.1 | 3 | 583 | |||
| XM_006712807.2 | XP_006712870.1 | 3 | 583 | |||
| XM_017005121.1 | XP_016860610.1 | 3 | 129 | |||
| GenBank mRNA | AK027587.1 | - | 3 | 611 | ||
| AK290294.1 | - | 3 | 272 | |||
| AY429534.1 | - | 3 | 129 | |||
| AY429535.1 | - | 3 | 129 | |||
| BC115373.1 | - | 3 | 430 | 40026807 | ||

TMEM87B
Genomic Map