siRNA Details
| Gene Symbol: FAM222A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032829.2 | NP_116218.2 | 3 | 2768 | ||
| XM_006719654.2 | XP_006719717.1 | 3 | 2354 | |||
| XM_017020055.1 | XP_016875544.1 | 3 | 2738 | |||
| GenBank mRNA | AK027627.1 | - | 3 | 2372 | ||
| AK074806.1 | - | 1 | 2027 | |||
| BC007844.2 | - | 2 | 833 | |||
| BC020193.1 | - | 3 | 2765 | |||
| BC037221.1 | - | 2 | 2243 | 5197135 | ||

FAM222A
Genomic Map