siRNA Details
| Gene Symbol: UTP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318391.1 | NP_001305320.1 | 11 | 1300 | ||
| NM_032830.2 | NP_116219.2 | 11 | 1354 | |||
| XM_005256205.3 | XP_005256262.2 | 7 | 885 | |||
| GenBank mRNA | AB075868.2 | - | 11 | 1292 | ||
| AK027419.1 | - | 8 | 909 | |||
| AK027634.1 | - | 11 | 1306 | |||
| AK027675.1 | - | 11 | 1303 | |||
| BC000167.2 | - | 7 | 799 | |||
| BC009348.2 | - | 11 | 1286 | 4131625 | ||
| BX647265.1 | - | 11 | 1268 | |||

UTP4
Genomic Map