siRNA Details
| Gene Symbol: TMEM209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032842.3 | NP_116231.2 | 10 | 1318 | ||
| XM_005250655.1 | XP_005250712.1 | 10 | 1608 | |||
| XM_011516646.2 | XP_011514948.1 | 10 | 1458 | |||
| GenBank mRNA | AK027709.1 | - | 10 | 1228 | ||
| AK074732.1 | - | 10 | 1251 | |||
| BC045618.1 | - | 10 | 1226 | 5294911 | ||

TMEM209
Genomic Map