siRNA Details
| Gene Symbol: TMEM209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301163.1 | NP_001288092.1 | 7 | 939 | ||
| NM_032842.3 | NP_116231.2 | 7 | 939 | |||
| XM_005250655.1 | XP_005250712.1 | 7 | 1229 | |||
| XM_011516646.2 | XP_011514948.1 | 7 | 1079 | |||
| GenBank mRNA | AK027709.1 | - | 7 | 849 | ||
| AK074732.1 | - | 7 | 872 | |||
| BC045618.1 | - | 7 | 847 | 5294911 | ||
| BC057784.1 | - | 7 | 844 | 5313685 | ||

TMEM209
Genomic Map