siRNA Details
| Gene Symbol: FIBCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145106.1 | NP_001138578.1 | 8 | 1288 | ||
| NM_032843.4 | NP_116232.3 | 7 | 1426 | |||
| GenBank mRNA | AK292489.1 | - | 7 | 1195 | ||
| AY358311.1 | - | 7 | 1183 | |||
| BC032953.2 | - | 7 | 1424 | 4811679 | ||
| BC112892.1 | - | 7 | 1176 | |||

FIBCD1
Genomic Map