siRNA Details
| Gene Symbol: CASP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004346.3 | NP_004337.2 | 4 | 359 | ||
| NM_032991.2 | NP_116786.1 | 3 | 192 | |||
| XM_011532301.1 | XP_011530603.1 | 4 | 293 | |||
| GenBank mRNA | AK291337.1 | - | 4 | 330 | ||
| AU127028.1 | - | 3 | 192 | |||
| AU133051.1 | - | 4 | 331 | |||
| BC016926.2 | - | 4 | 311 | 4419175 | ||
| BX647609.1 | - | 4 | 431 | |||
| U13737.1 | - | 4 | 320 | |||
| U13738.1 | - | 3 | 159 | |||
| U26943.1 | - | 2 | 96 | |||

CASP3
Genomic Map