siRNA Details
| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032268.4 | NP_115644.1 | 2, 3 | 1182 | ||
| XM_017023793.1 | XP_016879282.1 | 2, 3 | 1156 | |||
| GenBank mRNA | AF378524.1 | - | 2, 3 | 854 | ||
| AK294434.1 | - | 2, 3 | 854 | |||
| AL136903.1 | - | 2, 3 | 847 | |||
| BC007235.1 | - | 2, 3 | 854 | 2823677 | ||

ZNRF1
Genomic Map