siRNA Details
| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032268.4 | NP_115644.1 | 3 | 1269 | ||
| XM_017023793.1 | XP_016879282.1 | 3 | 1243 | |||
| GenBank mRNA | AF378524.1 | - | 3 | 941 | ||
| AK094870.1 | - | 1 | 80 | |||
| AK128349.1 | - | 1 | 818 | |||
| AK294434.1 | - | 3 | 941 | |||
| AL136903.1 | - | 3 | 934 | |||
| AL834440.1 | - | 4 | 746 | |||
| BC007235.1 | - | 3 | 941 | 2823677 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_133206, NM_001168621, NM_001168622, NM_001168623, AK216619, and others.

ZNRF1
Genomic Map