siRNA Details
| Gene Symbol: CORO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032854.3 | NP_116243.2 | 9 | 1166 | ||
| XM_005258047.3 | XP_005258104.1 | 10 | 1448 | |||
| XM_005258048.4 | XP_005258105.2 | 10 | 1385 | |||
| XM_005258056.4 | XP_005258113.1 | 5 | 1265 | |||
| XM_011525385.2 | XP_011523687.1 | 10 | 1385 | |||
| XM_011525386.2 | XP_011523688.1 | 9 | 1280 | |||
| XM_011525387.2 | XP_011523689.1 | 9 | 1262 | |||
| XM_011525388.2 | XP_011523690.1 | 10 | 1381 | |||
| XM_011525390.2 | XP_011523692.1 | 6 | 1128 | |||
| XM_011525391.2 | XP_011523693.1 | 6 | 1106 | |||
| XM_017025235.1 | XP_016880724.1 | 9 | 1329 | |||
| GenBank mRNA | AF193039.1 | - | 4 | 367 | ||
| AF447884.1 | - | 6 | 1150 | |||
| AF447885.1 | - | 4 | 836 | |||
| AK027777.1 | - | 4 | 838 | |||
| AK092430.1 | - | 7 | 1081 | |||
| AK094683.1 | - | 5 | 1988 | |||
| AK096431.1 | - | 9 | 1071 | |||
| AL833952.1 | - | 7 | 2557 | |||
| AY545070.1 | - | 4 | 606 | |||
| BC048209.1 | - | 6 | 580 | |||
| BC064514.1 | - | 6 | 580 | 3678185 | ||

CORO6
Genomic Map