siRNA Details
| Gene Symbol: LTV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032860.3 | NP_116249.2 | 10 | 1269 | ||
| GenBank mRNA | AK027815.1 | - | 8 | 1018 | ||
| AK091447.1 | - | 5 | 4318 | |||
| BC009855.2 | - | 10 | 1269 | 3926227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LTV1| Gene Symbol: LTV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032860.3 | NP_116249.2 | 10 | 1269 | ||
| GenBank mRNA | AK027815.1 | - | 8 | 1018 | ||
| AK091447.1 | - | 5 | 4318 | |||
| BC009855.2 | - | 10 | 1269 | 3926227 | ||