siRNA Details
Gene Symbol: MICALCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032867.2 | NP_116256.2 | 9 | 2311 | ||
GenBank mRNA | AK027872.1 | - | 7 | 1638 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MICALCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032867.2 | NP_116256.2 | 9 | 2311 | ||
GenBank mRNA | AK027872.1 | - | 7 | 1638 |