siRNA Details
| Gene Symbol: CCDC183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039374.4 | NP_001034463.4 | 9 | 939 | ||
| GenBank mRNA | AK090683.1 | - | 5 | 515 | ||
| AK092639.1 | - | 6 | 1197 | |||
| BC064582.1 | - | 9 | 925 | |||
| BC137317.1 | - | 9 | 914 | 9021314 | ||
| BC137318.1 | - | 9 | 914 | 9021315 | ||

CCDC183
Genomic Map