siRNA Details
| Gene Symbol: FBXL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184906.1 | NP_001171835.1 | 12 | 1120 | ||
| NM_032875.2 | NP_116264.2 | 13 | 1216 | |||
| XM_005257746.3 | XP_005257803.2 | 13 | 1407 | |||
| XM_005257747.3 | XP_005257804.2 | 12 | 1188 | |||
| GenBank mRNA | AK123048.1 | - | 13 | 1217 | ||
| AK291844.1 | - | 13 | 1227 | |||
| AK312612.1 | - | 13 | 1197 | |||
| BC007557.1 | - | 13 | 1183 | 2987858 | ||
| DQ223959.1 | - | 12 | 1087 | |||

FBXL20
Genomic Map