siRNA Details
Gene Symbol: PNMA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032882.5 | NP_116271.3 | 2 | 2237 | ||
GenBank mRNA | AK055322.1 | - | 2 | 2169 | ||
BC007631.1 | - | 1 | 1969 | 3507248 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PNMA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032882.5 | NP_116271.3 | 2 | 2237 | ||
GenBank mRNA | AK055322.1 | - | 2 | 2169 | ||
BC007631.1 | - | 1 | 1969 | 3507248 |