siRNA Details
| Gene Symbol: ATG4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281504.1 | NP_001268433.1 | 5 | 1081 | ||
| NM_032885.5 | NP_116274.3 | 5 | 1007 | |||
| NR_104024.1 | 5 | 963 | ||||
| NR_104025.1 | 4 | 730 | ||||
| GenBank mRNA | AJ312332.1 | - | 5 | 823 | ||
| AK056210.1 | - | 5 | 982 | |||
| AK223383.1 | - | 5 | 932 | |||
| AK294676.1 | - | 5 | 1081 | |||
| AK314712.1 | - | 5 | 942 | |||
| BC007639.1 | - | 4 | 653 | |||
| BC068992.1 | - | 5 | 983 | 6578102 | ||

ATG4D
Genomic Map