siRNA Details
| Gene Symbol: FRMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286490.1 | NP_001273419.1 | 7, 8 | 741 | ||
| NM_001286491.1 | NP_001273420.1 | 8, 9 | 1199 | |||
| NM_001322949.1 | NP_001309878.1 | 6, 7 | 791 | |||
| NM_001322950.1 | NP_001309879.1 | 6, 7 | 791 | |||
| NM_001322951.1 | NP_001309880.1 | 5, 6 | 686 | |||
| NM_032892.4 | NP_116281.2 | 6, 7 | 791 | |||
| NR_104455.1 | 7, 8 | 1094 | ||||
| XM_005254730.3 | XP_005254787.1 | 6, 7 | 696 | |||
| GenBank mRNA | AK090572.1 | - | 6, 7 | 723 | ||
| AK123018.1 | - | 7, 8 | 741 | |||
| BC043486.2 | - | 8, 9 | 946 | |||
| BC053647.1 | - | 6, 7 | 728 | 5578205 | ||

FRMD5
Genomic Map