siRNA Details
| Gene Symbol: TMEM141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032928.3 | NP_116317.1 | 3 | 194 | ||
| GenBank mRNA | BC007834.2 | - | 3 | 164 | 4100086 | |
| CB107060.1 | - | 3 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM141| Gene Symbol: TMEM141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032928.3 | NP_116317.1 | 3 | 194 | ||
| GenBank mRNA | BC007834.2 | - | 3 | 164 | 4100086 | |
| CB107060.1 | - | 3 | 194 | |||