siRNA Details
| Gene Symbol: C11orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032930.2 | NP_116319.2 | 4 | 378 | ||
| XM_005271713.3 | XP_005271770.1 | 4 | 406 | |||
| XM_017018454.1 | XP_016873943.1 | 4 | 407 | |||
| GenBank mRNA | AK094851.1 | - | 4 | 358 | ||
| BC006128.1 | - | 3 | 582 | 3622924 | ||
| BG700929.1 | - | 4 | 382 | |||

C11orf70
Genomic Map