siRNA Details
| Gene Symbol: TMEM241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318834.1 | NP_001305763.1 | 10, 11 | 761 | ||
| NM_032933.5 | NP_116322.3 | 9, 10 | 646 | |||
| NR_134875.1 | 8, 9 | 597 | ||||
| XM_011526233.2 | XP_011524535.1 | 9, 10 | 636 | |||
| XM_017026043.1 | XP_016881532.1 | 9, 10 | 636 | |||
| XM_017026044.1 | XP_016881533.1 | 9, 10 | 636 | |||
| XM_017026045.1 | XP_016881534.1 | 8, 9 | 560 | |||
| XM_017026046.1 | XP_016881535.1 | 9, 10 | 642 | |||
| XM_017026047.1 | XP_016881536.1 | 9, 10 | 566 | |||
| XM_017026049.1 | XP_016881538.1 | 9, 10 | 642 | |||
| XM_017026050.1 | XP_016881539.1 | 9, 10 | 664 | |||
| XM_017026051.1 | XP_016881540.1 | 9, 10 | 566 | |||
| GenBank mRNA | AB253738.1 | - | 9, 10 | 546 | ||
| AB253739.1 | - | 10, 11 | 661 | |||
| AK126247.1 | - | 8, 9 | 589 | |||
| AK294916.1 | - | 9, 10 | 642 | |||
| AK314123.1 | - | 1, 2 | 48 | |||
| BC054507.1 | - | 3, 4 | 143 | |||
| BC114510.1 | - | 8, 9 | 533 | 40023922 | ||
| BC114561.1 | - | 9, 10 | 582 | 40023923 | ||
| DA385968.1 | - | 8, 9 | 561 | |||

TMEM241
Genomic Map