siRNA Details
| Gene Symbol: TMEM241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318834.1 | NP_001305763.1 | 4 | 371 | ||
| NM_032933.5 | NP_116322.3 | 4 | 371 | |||
| NR_134875.1 | 4 | 371 | ||||
| NR_134876.1 | 3 | 293 | ||||
| NR_134877.1 | 4 | 371 | ||||
| XM_011526233.2 | XP_011524535.1 | 4 | 361 | |||
| XM_017026043.1 | XP_016881532.1 | 4 | 361 | |||
| XM_017026044.1 | XP_016881533.1 | 4 | 361 | |||
| XM_017026045.1 | XP_016881534.1 | 4 | 363 | |||
| XM_017026047.1 | XP_016881536.1 | 4 | 254 | |||
| XM_017026050.1 | XP_016881539.1 | 3 | 274 | |||
| XM_017026051.1 | XP_016881540.1 | 4 | 254 | |||
| GenBank mRNA | AB253738.1 | - | 4 | 271 | ||
| AB253739.1 | - | 4 | 271 | |||
| AK123623.1 | - | 3 | 283 | |||
| AK126247.1 | - | 4 | 363 | |||
| AK303553.1 | - | 4 | 255 | |||
| BC114510.1 | - | 4 | 307 | 40023922 | ||
| BC114561.1 | - | 4 | 307 | 40023923 | ||
| DA385968.1 | - | 3 | 286 | |||
| DC334544.1 | - | 4 | 373 | |||

TMEM241
Genomic Map