siRNA Details
| Gene Symbol: TMEM60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032936.3 | NP_116325.1 | 2 | 519 | ||
| GenBank mRNA | AF260336.1 | - | 2 | 520 | ||
| AK095664.1 | - | 3674 | ||||
| BC065930.1 | - | 2 | 443 | 6146561 | ||
| BX117806.1 | - | 2 | 519 | |||

TMEM60
Genomic Map