siRNA Details
Gene Symbol: KRTAP4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033060.2 | NP_149049.1 | 2 | 636 | ||
GenBank mRNA | AJ406942.1 | - | 2 | 636 | ||
BC065372.1 | - | 3 | 1133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033060.2 | NP_149049.1 | 2 | 636 | ||
GenBank mRNA | AJ406942.1 | - | 2 | 636 | ||
BC065372.1 | - | 3 | 1133 |