siRNA Details
| Gene Symbol: KRTAP4-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033188.3 | NP_149445.3 | 1 | 742 | ||
| GenBank mRNA | AJ406937.1 | - | 1 | 757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-5| Gene Symbol: KRTAP4-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033188.3 | NP_149445.3 | 1 | 742 | ||
| GenBank mRNA | AJ406937.1 | - | 1 | 757 | ||