siRNA Details
| Gene Symbol: KIAA1644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099294.1 | NP_001092764.1 | 3 | 327 | ||
| XM_005261790.3 | XP_005261847.1 | 3 | 425 | |||
| GenBank mRNA | BC104183.1 | - | 2 | 194 | 40030810 | |
| BC104184.1 | - | 2 | 194 | 40030814 | ||

KIAA1644
Genomic Map