siRNA Details
| Gene Symbol: SHANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033517.1 | NP_277052.1 | 22 | 5075 | ||
| GenBank mRNA | AB051437.1 | - | 2 | 2274 | ||
| AB569469.1 | - | 22 | 5075 | |||
| AK074038.1 | - | 2 | 2193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHANK3| Gene Symbol: SHANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033517.1 | NP_277052.1 | 22 | 5075 | ||
| GenBank mRNA | AB051437.1 | - | 2 | 2274 | ||
| AB569469.1 | - | 22 | 5075 | |||
| AK074038.1 | - | 2 | 2193 | |||