siRNA Details
| Gene Symbol: CCNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033031.2 | NP_149020.2 | 9 | 3850 | ||
| NM_033670.2 | NP_391990.1 | 5 | 538 | |||
| XM_017029913.1 | XP_016885402.1 | 11 | 4061 | |||
| XM_017029914.1 | XP_016885403.1 | 11 | 3991 | |||
| XM_017029915.1 | XP_016885404.1 | 12 | 4021 | |||
| XM_017029916.1 | XP_016885405.1 | 11 | 4058 | |||
| XM_017029917.1 | XP_016885406.1 | 10 | 4037 | |||
| XM_017029918.1 | XP_016885407.1 | 10 | 4034 | |||
| GenBank mRNA | AJ314764.1 | - | 9 | 3854 | ||
| AJ314765.1 | - | 5 | 542 | |||
| AJ314766.1 | - | 8 | 862 | |||
| AJ416458.1 | - | 9 | 3866 | |||
| AK292454.1 | - | 10 | 4023 | |||
| AY927611.1 | - | 2 | 715 | |||

CCNB3
Genomic Map