siRNA Details
| Gene Symbol: DCLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033403.1 | NP_208382.1 | 4 | 2180 | ||
| XM_011534167.2 | XP_011532469.1 | 4 | 2139 | |||
| GenBank mRNA | AB051552.1 | - | 3 | 1570 | ||
| AK122988.1 | - | 3 | 1499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCLK3| Gene Symbol: DCLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033403.1 | NP_208382.1 | 4 | 2180 | ||
| XM_011534167.2 | XP_011532469.1 | 4 | 2139 | |||
| GenBank mRNA | AB051552.1 | - | 3 | 1570 | ||
| AK122988.1 | - | 3 | 1499 | |||