siRNA Details
| Gene Symbol: KIAA1755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029864.1 | NP_001025035.1 | 3 | 996 | ||
| GenBank mRNA | AB051542.1 | - | 1 | 454 | ||
| AK096202.1 | - | 3 | 996 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1755| Gene Symbol: KIAA1755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029864.1 | NP_001025035.1 | 3 | 996 | ||
| GenBank mRNA | AB051542.1 | - | 1 | 454 | ||
| AK096202.1 | - | 3 | 996 | |||