siRNA Details
| Gene Symbol: UNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080419.2 | NP_001073888.2 | 12 | 1652 | ||
| XM_006722145.3 | XP_006722208.1 | 15 | 5997 | |||
| XM_006722148.2 | XP_006722211.1 | 13 | 1793 | |||
| XM_011525400.1 | XP_011523702.1 | 13 | 1721 | |||
| XM_011525401.2 | XP_011523703.1 | 7 | 809 | |||
| XM_017025247.1 | XP_016880736.1 | 15 | 6033 | |||
| XM_017025248.1 | XP_016880737.1 | 14 | 5935 | |||
| XM_017025249.1 | XP_016880738.1 | 14 | 2078 | |||
| XM_017025250.1 | XP_016880739.1 | 13 | 1980 | |||
| GenBank mRNA | AB051540.1 | - | 12 | 1635 | ||
| BC053362.1 | - | 12 | 1635 | |||

UNK
Genomic Map