siRNA Details
| Gene Symbol: CCDC65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286957.1 | NP_001273886.1 | 8 | 1520 | ||
| NM_033124.4 | NP_149115.2 | 8 | 1560 | |||
| GenBank mRNA | AF382188.1 | - | 8 | 1525 | ||
| AK093051.1 | - | 8 | 1533 | |||
| AK098529.1 | - | 5 | 793 | |||
| AK302082.1 | - | 8 | 1497 | |||
| AK314623.1 | - | 8 | 1505 | |||
| BC039317.1 | - | 8 | 1499 | 5271866 | ||

CCDC65
Genomic Map