siRNA Details
| Gene Symbol: TSLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033035.4 | NP_149024.1 | 4 | 596 | ||
| NM_138551.4 | NP_612561.2 | 2 | 378 | |||
| NR_045089.1 | 5 | 1801 | ||||
| XM_011543698.1 | XP_011542000.1 | 3 | 200 | |||
| GenBank mRNA | AF338732.1 | - | 4 | 430 | ||
| AL833421.1 | - | 2 | 358 | |||
| AY037115.1 | - | 4 | 596 | |||
| BC016720.1 | - | 2 | 311 | |||
| BC022894.1 | - | 2 | 350 | |||
| BC040592.1 | - | 5 | 1802 | 5269764 | ||
| BF245025.1 | - | 2 | 366 | |||
| BU571002.1 | - | 2 | 381 | |||
 Back To Top
 Back To Top
 TSLP
TSLP Genomic Map
Genomic Map Back To Top
 Back To Top