siRNA Details
| Gene Symbol: PSKH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033126.1 | NP_149117.1 | 2 | 588 | ||
| XM_017013929.1 | XP_016869418.1 | 4 | 945 | |||
| GenBank mRNA | AY037806.1 | - | 2 | 588 | ||
| BC126180.1 | - | 2 | 663 | 8991896 | ||
| BC126182.1 | - | 2 | 663 | 8991898 | ||

PSKH2
Genomic Map