siRNA Details
| Gene Symbol: SCRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033129.3 | NP_149120.1 | 2 | 1933 | ||
| GenBank mRNA | AY129025.1 | - | 2 | 1939 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCRT2| Gene Symbol: SCRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033129.3 | NP_149120.1 | 2 | 1933 | ||
| GenBank mRNA | AY129025.1 | - | 2 | 1939 | ||