siRNA Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033017.3 | NP_148977.2 | 7 | 1413 | ||
| NM_033091.2 | NP_149082.1 | 6 | 1335 | |||
| GenBank mRNA | AF220023.1 | - | 8 | 1350 | ||
| AF220024.1 | - | 6 | 1272 | |||
| AK092892.1 | - | 6 | 1297 | |||
| AK293565.1 | - | 8 | 1189 | |||

TRIM4
Genomic Map