siRNA Details
| Gene Symbol: NAV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024383.1 | NP_001019554.1 | 16 | 4291 | ||
| NM_014903.5 | NP_055718.4 | 16 | 4291 | |||
| XM_005269215.3 | XP_005269272.1 | 16 | 8292 | |||
| XM_011538940.2 | XP_011537242.1 | 16 | 8293 | |||
| XM_011538941.2 | XP_011537243.1 | 16 | 8292 | |||
| XM_017020164.1 | XP_016875653.1 | 16 | 8294 | |||
| XM_017020165.1 | XP_016875654.1 | 16 | 8294 | |||
| XM_017020166.1 | XP_016875655.1 | 16 | 4280 | |||
| XM_017020167.1 | XP_016875656.1 | 15 | 4211 | |||
| XM_017020171.1 | XP_016875660.1 | 8 | 7587 | |||
| XM_017020172.1 | XP_016875661.1 | 8 | 3225 | |||
| GenBank mRNA | AF397731.1 | - | 15 | 3875 | ||
| AJ488201.1 | - | 16 | 4163 | |||
| AK298525.1 | - | 1 | 47 | |||

NAV3
Genomic Map