siRNA Details
| Gene Symbol: AQP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080429.2 | NP_536354.2 | 6 | 1102 | ||
| XM_011510104.2 | XP_011508406.1 | 6 | 1162 | |||
| GenBank mRNA | AB066105.1 | - | 6 | 1102 | ||
| AF159174.1 | - | 5 | 1573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AQP10| Gene Symbol: AQP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080429.2 | NP_536354.2 | 6 | 1102 | ||
| XM_011510104.2 | XP_011508406.1 | 6 | 1162 | |||
| GenBank mRNA | AB066105.1 | - | 6 | 1102 | ||
| AF159174.1 | - | 5 | 1573 | |||