siRNA Details
| Gene Symbol: TPD52L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001874.2 | NP_001001874.1 | 1 | 375 | ||
| NM_001001875.3 | NP_001001875.1 | 1 | 375 | |||
| NM_033516.5 | NP_277051.3 | 1 | 375 | |||
| XM_017015280.1 | XP_016870769.1 | 1 | 363 | |||
| GenBank mRNA | AK057479.1 | - | 1 | 349 | ||
| AY032877.1 | - | 1 | 353 | |||
| BC017589.2 | - | 1 | 349 | 4828082 | ||
| BC033792.1 | - | 1 | 320 | 5163651 | ||
| BC042113.1 | - | 1 | 334 | 4838253 | ||
| BI463769.1 | - | 392 | ||||
| BP369721.1 | - | 1 | 375 | |||

TPD52L3
Genomic Map