siRNA Details
| Gene Symbol: C16orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142469.1 | NP_001135941.1 | 5 | 478 | ||
| NM_033201.2 | NP_149978.1 | 5 | 659 | |||
| XM_011522731.2 | XP_011521033.1 | 5 | 638 | |||
| XM_017023824.1 | XP_016879313.1 | 6 | 785 | |||
| GenBank mRNA | AK057180.1 | - | 5 | 659 | ||
| AK293431.1 | - | 5 | 565 | |||
| BC008967.2 | - | 5 | 589 | 3010666 | ||
| BC023603.2 | - | 5 | 478 | 4330351 | ||

C16orf45
Genomic Map