siRNA Details
| Gene Symbol: DPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141972.1 | NP_001135444.1 | 3 | 296 | ||
| NM_080650.3 | NP_542381.1 | 3 | 296 | |||
| XM_011522160.1 | XP_011520462.1 | 3 | 315 | |||
| XM_017022707.1 | XP_016878196.1 | 3 | 315 | |||
| XM_017022708.1 | XP_016878197.1 | 3 | 315 | |||
| XM_017022709.1 | XP_016878198.1 | 3 | 315 | |||
| GenBank mRNA | AK094007.1 | - | 3 | 256 | ||
| AK125017.1 | - | 3 | 285 | |||
| AL833427.1 | - | 3 | 257 | |||
| BC008485.2 | - | 2 | 212 | |||
| BC066652.1 | - | 3 | 243 | 6066017 | ||
| DB250468.1 | - | 3 | 296 | |||

DPH6
Genomic Map