siRNA Details
| Gene Symbol: FLJ20021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033874.1 | 2 | 598 | |||
| GenBank mRNA | AK000028.1 | - | 2 | 594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ20021| Gene Symbol: FLJ20021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033874.1 | 2 | 598 | |||
| GenBank mRNA | AK000028.1 | - | 2 | 594 | ||