siRNA Details
| Gene Symbol: CCDC120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163323.2 | NP_001156795.1 | 10 | 2262 | ||
| NM_001271835.1 | NP_001258764.1 | 10 | 2262 | |||
| NM_001271836.1 | NP_001258765.1 | 10 | 2614 | |||
| NM_033626.3 | NP_296375.1 | 10 | 2203 | |||
| GenBank mRNA | AK056290.1 | - | 1 | 799 | ||
| AK125424.1 | - | 1 | 1797 | |||
| AK291370.1 | - | 10 | 2474 | |||
| AK300366.1 | - | 10 | 2213 | |||
| AK315837.1 | - | 10 | 2229 | |||
| BC008769.2 | - | 10 | 2184 | 3139519 | ||

CCDC120
Genomic Map