siRNA Details
| Gene Symbol: PHLDB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134437.1 | NP_001127909.1 | 3 | 1195 | ||
| NM_001134438.1 | NP_001127910.1 | 2 | 1253 | |||
| NM_001134439.1 | NP_001127911.1 | 2 | 1014 | |||
| NM_145753.2 | NP_665696.1 | 2 | 1253 | |||
| GenBank mRNA | AB209903.1 | - | 2 | 895 | ||
| AF506820.1 | - | 1 | 957 | |||
| AJ496194.1 | - | 1 | 868 | |||
| AK092996.1 | - | 2 | 939 | |||
| AK096151.1 | - | 2 | 1195 | |||
| AK296841.1 | - | 3 | 1195 | |||
| AK307491.1 | - | 3 | 1157 | |||
| AK315050.1 | - | 2 | 1165 | |||
| AL832205.1 | - | 2 | 976 | |||
| AL832476.1 | - | 2 | 942 | |||
| BC038806.1 | - | 2 | 985 | |||
| BC069194.1 | - | 2 | 1142 | |||
| BC142678.1 | - | 1 | 860 | 40148225 | ||
| BC150210.1 | - | 1 | 860 | 40148898 | ||
| CR749654.1 | - | 3 | 1178 | |||

PHLDB2
Genomic Map